Gene Transcripts
- Gene Symbol
- Apoe
- Entrez Gene ID
- 11816
- Gene Name
- apolipoprotein E
- Gene Aliases
- AI255918, Apo-E
- Chromosome Location
- Chr. 7: 19696244 - 19699166 on Build GRCm38
- UniGene ID
- Mm.305152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK010261.1 |
- |
3, 4 |
276 |
|
AK131624.1 |
- |
3, 4 |
276 |
|
AK134921.1 |
- |
3, 4 |
276 |
|
AK149111.1 |
- |
3, 4 |
278 |
|
AK149568.1 |
- |
3, 4 |
258 |
|
AK150834.1 |
- |
3, 4 |
278 |
|
AK159105.1 |
- |
3, 4 |
276 |
|
AK159424.1 |
- |
3, 4 |
278 |
|
AK159517.1 |
- |
3, 4 |
276 |
|
BC028816.1 |
- |
3, 4 |
255 |
5136415 |
BC044785.1 |
- |
2, 3 |
247 |
|
BC083351.1 |
- |
2, 3 |
245 |
6818357 |
CT010212.1 |
- |
2, 3 |
210 |
|
CT010356.1 |
- |
2, 3 |
210 |
|
DN173921.1 |
- |
2, 3 |
378 |
|
M12414.1 |
- |
2, 3 |
210 |
|
M73490.1 |
- |
1, 2 |
137 |
|
View Full Product Details