Gene Transcripts

Gene Symbol
HDAC2
Entrez Gene ID
3066
Gene Name
histone deacetylase 2
Gene Aliases
HD2, RPD3, YAF1
Chromosome Location
Chr. 6: 113936156 - 113971195 on Build GRCh38
UniGene ID
Hs.3352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001527.3 NP_001518.3 7, 8 1011
NR_033441.1 8, 9 1039
NR_073443.1 7, 8 917
XM_011535788.1 XP_011534090.1 8, 9 763
XM_017010799.1 XP_016866288.1 8, 9 985
GenBank mRNA AK092156.1 - 7, 8 917
AK097376.1 - 8, 9 973
AK296856.1 - 7, 8 1011
BC031055.1 - 7, 8 975
U31814.1 - 7, 8 928

View Full Product Details