Gene Transcripts

Gene Symbol
Emb
Entrez Gene ID
13723
Gene Name
embigin
Gene Aliases
AL022799, Gp70
Chromosome Location
Chr. 13: 117220573 - 117274415 on Build GRCm38
UniGene ID
Mm.274926
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010330.4 NP_034460.3 4 694
GenBank mRNA AK079570.1 - 4 636
AK088480.1 - 4 522
AK138271.1 - 4 643
AK148449.1 - 4 522
AK148540.1 - 4 522
AK150625.1 - 4 523
AK161679.1 - 4 522
AK166602.1 - 4 522
AK167997.1 - 4 554
AK200349.1 - 2 191
BC014858.1 - 4 513 4500922
J03535.2 - 4 521

View Full Product Details