Gene Transcripts

Gene Symbol
Emx2
Entrez Gene ID
13797
Gene Name
empty spiracles homeobox 2
Gene Aliases
Pdo
Chromosome Location
Chr. 19: 59458116 - 59465357 on Build GRCm38
UniGene ID
Mm.245394
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010132.2 NP_034262.2 3 1189
GenBank mRNA AK132093.1 - 3 977
AY117415.1 - 3 1507

View Full Product Details