Gene Transcripts

Gene Symbol
Lhx6
Entrez Gene ID
16874
Gene Name
LIM homeobox protein 6
Gene Aliases
Lhx6.1
Chromosome Location
Chr. 2: 36081950 - 36105997 on Build GRCm38
UniGene ID
Mm.12881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083125.1 NP_001076594.1 8 1110
NM_001083126.1 NP_001076595.1 7 920
NM_001083127.1 NP_001076596.1 7 920
NM_008500.2 NP_032526.2 8 1110
GenBank mRNA AB031039.1 - 7 920
AB031040.1 - 7 920
AJ000337.1 - 8 952
AK140987.1 - 8 1112
AK149585.1 - 2 114
BC065077.1 - 7 854 6816328

View Full Product Details