Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001293648.1 | NP_001280577.1 | 13 | 2183 | |
NM_019676.3 | NP_062650.1 | 13 | 2134 | ||
GenBank mRNA | AF133125.1 | - | 13 | 2116 | |
AK028749.1 | - | 13 | 2135 | ||
AK082890.1 | - | 13 | 2126 | ||
BC015249.1 | - | 13 | 2146 | 4241580 | |
BC025798.1 | - | 13 | 2110 | 5068610 | |
JN635596.1 | - | 13 | 2183 | ||
U85711.1 | - | 13 | 2128 |