Gene Transcripts
- Gene Symbol
- Rps3a1
- Entrez Gene ID
- 20091
- Gene Name
- ribosomal protein S3A1
- Gene Aliases
- Rps3a
- Chromosome Location
- Chr. 3: 86137940 - 86142668 on Build GRCm38
- UniGene ID
- Mm.391785
- Species
- Mus musculus
- Gene Symbol
- Sox10
- Entrez Gene ID
- 20665
- Gene Name
- SRY (sex determining region Y)-box 10
- Gene Aliases
- Dom, Sox21
- Chromosome Location
- Chr. 15: 79154909 - 79165235 on Build GRCm38
- UniGene ID
- Mm.276739
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK189901.1 |
- |
|
275 |
|
View Full Product Details