Gene Transcripts
- Gene Symbol
- IL12RB1
- Entrez Gene ID
- 3594
- Gene Name
- interleukin 12 receptor subunit beta 1
- Gene Aliases
- CD212, IL-12R-BETA1, IL12RB, IMD30
- Chromosome Location
- Chr. 19: 18058994 - 18099027 on Build GRCh38
- UniGene ID
- Hs.567294
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290023.1 |
NP_001276952.1
|
4, 5 |
581 |
|
NM_001290024.1 |
NP_001276953.1
|
5, 6 |
572 |
|
NM_005535.2 |
NP_005526.1
|
4, 5 |
581 |
|
NM_153701.2 |
NP_714912.1
|
4, 5 |
581 |
|
XM_006722741.3 |
XP_006722804.2
|
5, 6 |
774 |
|
XM_011527966.2 |
XP_011526268.1
|
5, 6 |
795 |
|
XM_011527967.2 |
XP_011526269.1
|
5, 6 |
781 |
|
XM_011527968.2 |
XP_011526270.1
|
5, 6 |
789 |
|
XM_011527969.2 |
XP_011526271.1
|
5, 6 |
775 |
|
XM_011527970.2 |
XP_011526272.1
|
5, 6 |
794 |
|
XM_011527971.2 |
XP_011526273.1
|
5, 6 |
792 |
|
XM_011527972.2 |
XP_011526274.1
|
5, 6 |
792 |
|
XM_011527973.2 |
XP_011526275.1
|
4, 5 |
586 |
|
XM_011527974.2 |
XP_011526276.1
|
4, 5 |
574 |
|
XM_011527975.2 |
XP_011526277.1
|
5, 6 |
772 |
|
XM_011527976.2 |
XP_011526278.1
|
5, 6 |
792 |
|
XM_011527977.2 |
XP_011526279.1
|
5, 6 |
765 |
|
GenBank mRNA |
AK225919.1 |
- |
4, 5 |
512 |
|
AK290423.1 |
- |
4, 5 |
509 |
|
BC029121.1 |
- |
4, 5 |
470 |
|
BC137404.1 |
- |
4, 5 |
426 |
9021402 |
BC137406.1 |
- |
4, 5 |
426 |
9021404 |
BX647221.1 |
- |
5, 6 |
572 |
|
U03187.1 |
- |
4, 5 |
465 |
|
View Full Product Details