Gene Transcripts
- Gene Symbol
- Tnfsf11
- Entrez Gene ID
- 21943
- Gene Name
- tumor necrosis factor (ligand) superfamily, member 11
- Gene Aliases
- Ly109l, ODF, OPG, OPGL, RANKL, Trance
- Chromosome Location
- Chr. 14: 78277446 - 78308043 on Build GRCm38
- UniGene ID
- Mm.249221
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011613.3 |
NP_035743.2
|
1, 2 |
387 |
|
GenBank mRNA |
AB008426.1 |
- |
1, 2 |
233 |
|
AB032771.1 |
- |
2, 3 |
146 |
|
AB036798.1 |
- |
1, 2 |
233 |
|
AF013170.1 |
- |
1, 2 |
374 |
|
AF019048.1 |
- |
1, 2 |
369 |
|
AF053713.1 |
- |
1, 2 |
402 |
|
AK159498.1 |
- |
1, 2 |
388 |
|
AK159997.1 |
- |
1, 2 |
388 |
|
BC125603.1 |
- |
1, 2 |
290 |
40130118 |
BC131970.1 |
- |
1, 2 |
290 |
40130247 |
View Full Product Details