Gene Transcripts

Gene Symbol
Mapk14
Entrez Gene ID
26416
Gene Name
mitogen-activated protein kinase 14
Gene Aliases
CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha
Chromosome Location
Chr. 17: 28691342 - 28748405 on Build GRCm38
UniGene ID
Mm.311337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168508.1 NP_001161980.1 6 870
NM_001168513.1 NP_001161985.1 6 471
NM_001168514.1 NP_001161986.1 7 915
NM_011951.3 NP_036081.1 6 870
GenBank mRNA AF128892.1 - 6 768
AK089059.1 - 6 784
AK133684.1 - 6 783
AK151348.1 - 6 472
AK153025.1 - 6 472
AK199780.1 - 212
BC012235.1 - 6 870 4195842
BC018227.1 - 7 812
BC085303.1 - 6 846 5389457
CX568915.1 - 5 264
D83073.1 - 6 701
U10871.1 - 6 768
X65067.1 - 1 13

View Full Product Details