Gene Transcripts
- Gene Symbol
- LAMA4
- Entrez Gene ID
- 3910
- Gene Name
- laminin subunit alpha 4
- Gene Aliases
- CMD1JJ, LAMA3, LAMA4*-1
- Chromosome Location
- Chr. 6: 112107931 - 112254722 on Build GRCh38
- UniGene ID
- Hs.654572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001105206.2 |
NP_001098676.2
|
35, 36 |
5373 |
|
NM_001105207.2 |
NP_001098677.2
|
35, 36 |
5369 |
|
NM_002290.4 |
NP_002281.3
|
35, 36 |
5352 |
|
XM_005266983.3 |
XP_005267040.2
|
35, 36 |
5246 |
|
XM_005266984.3 |
XP_005267041.2
|
35, 36 |
5243 |
|
XM_017010854.1 |
XP_016866343.1
|
35, 36 |
5222 |
|
GenBank mRNA |
AB210027.1 |
- |
35, 36 |
5225 |
|
BC066552.1 |
- |
35, 36 |
5365 |
6164247 |
S78569.1 |
- |
35, 36 |
5144 |
|
X70904.1 |
- |
5, 6 |
748 |
|
X76939.1 |
- |
29, 30 |
4249 |
|
X91171.1 |
- |
35, 36 |
5237 |
|
View Full Product Details