Gene Transcripts

Gene Symbol
Gmpr
Entrez Gene ID
66355
Gene Name
guanosine monophosphate reductase
Gene Aliases
2310004P21Rik, AV028449
Chromosome Location
Chr. 13: 45507429 - 45546386 on Build GRCm38
UniGene ID
Mm.290802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK002326.1 - 2 293
AK009159.1 - 2 304
AK084508.1 - 2 221
BC010827.1 - 2 275 4219628
BY274415.1 - 2 305

View Full Product Details