Gene Transcripts
- Gene Symbol
- Cox20
- Entrez Gene ID
- 66359
- Gene Name
- COX20 Cox2 chaperone
- Gene Aliases
- 2310005N03Rik, Fam36a
- Chromosome Location
- Chr. 1: 178319153 - 178322693 on Build GRCm38
- UniGene ID
- Mm.489720
- Species
- Mus musculus
- Gene Symbol
- AI503316
- Entrez Gene ID
- 105860
- Gene Name
- expressed sequence AI503316
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.453372
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK153188.1 |
- |
1 |
1151 |
|
View Full Product Details