Gene Transcripts

Gene Symbol
Fam217b
Entrez Gene ID
71532
Gene Name
family with sequence similarity 217, member B
Gene Aliases
9030418K01Rik, AI851938
Chromosome Location
Chr. 2: 178414534 - 178422181 on Build GRCm38
UniGene ID
Mm.220761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081289.1 NP_001074758.1 4 407
GenBank mRNA AK155442.1 - 4 408

View Full Product Details