Gene Transcripts

Gene Symbol
Phactr4
Entrez Gene ID
100169
Gene Name
phosphatase and actin regulator 4
Gene Aliases
3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120
Chromosome Location
Chr. 4: 132355922 - 132422446 on Build GRCm38
UniGene ID
Mm.158361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175306.4 NP_780515.2 4 347
GenBank mRNA AK049209.1 - 4 310
BC075672.1 - 4 309 30549496
BC096033.1 - 4 266 6854767

View Full Product Details