Gene Transcripts

Gene Symbol
Nudt10
Entrez Gene ID
102954
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 10
Gene Aliases
AI853080, Dipp3a
Chromosome Location
Chr. X: 6168696 - 6173044 on Build GRCm38
UniGene ID
Mm.426101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031664.1 NP_001026834.1 2 1394
GenBank mRNA BC055771.1 - 2 1351

View Full Product Details