Gene Transcripts
- Gene Symbol
- Slc16a6
- Entrez Gene ID
- 104681
- Gene Name
- solute carrier family 16 (monocarboxylic acid transporters), member 6
- Gene Aliases
- AW743111, ESTM12, MCT 6, MCT 7, MCT6
- Chromosome Location
- Chr. 11: 109450855 - 109473596 on Build GRCm38
- UniGene ID
- Mm.265874
- Species
- Mus musculus
- Gene Symbol
- LOC100503496
- Entrez Gene ID
- 100503496
- Gene Name
- uncharacterized LOC100503496
- Gene Aliases
- Chromosome Location
- Chr. 11: 109440078 - 109453359 on Build GRCm38
- UniGene ID
- Mm.265874
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BG090538.1 |
- |
1 |
146 |
|
View Full Product Details