Gene Transcripts

Gene Symbol
4930555G01Rik
Entrez Gene ID
108978
Gene Name
RIKEN cDNA 4930555G01 gene
Gene Aliases
Chromosome Location
Chr. 14: 5050629 - 5059243 on Build GRCm38
UniGene ID
Mm.396152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175393.4 NP_780602.2 5 1495
GenBank mRNA AK076920.1 - 5 1496

View Full Product Details