Gene Transcripts
- Gene Symbol
- 4930555G01Rik
- Entrez Gene ID
- 108978
- Gene Name
- RIKEN cDNA 4930555G01 gene
- Gene Aliases
- Chromosome Location
- Chr. 14: 5050629 - 5059243 on Build GRCm38
- UniGene ID
- Mm.396152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175393.4 |
NP_780602.2
|
1, 2 |
215 |
|
GenBank mRNA |
AK076920.1 |
- |
1, 2 |
216 |
|
View Full Product Details