Gene Transcripts
- Gene Symbol
- Olfr821
- Entrez Gene ID
- 258772
- Gene Name
- olfactory receptor 821
- Gene Aliases
- MOR109-1
- Chromosome Location
- Chr. 10: 130030390 - 130034560 on Build GRCm38
- UniGene ID
- Mm.391459
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146776.1 |
NP_666987.1
|
1 |
75 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB174445.1 |
- |
|
584 |
|
Chr. - - [-] |
View Full Product Details