Gene Transcripts

Gene Symbol
CENPB
Entrez Gene ID
1059
Gene Name
centromere protein B
Gene Aliases
Chromosome Location
Chr. 20: 3783851 - 3786690 on Build GRCh38
UniGene ID
Hs.516855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001810.5 NP_001801.1 1 1568
GenBank mRNA BC053847.1 - 1 1568
X05299.1 - 1 1346

View Full Product Details