Gene Transcripts
- Gene Symbol
- FMR1
- Entrez Gene ID
- 2332
- Gene Name
- fragile X mental retardation 1
- Gene Aliases
- FMRP, FRAXA, POF, POF1
- Chromosome Location
- Chr. X: 147911951 - 147951127 on Build GRCh38
- UniGene ID
- Hs.103183
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001185075.1 |
NP_001172004.1
|
3, 4 |
435 |
|
NM_001185076.1 |
NP_001172005.1
|
3, 4 |
435 |
|
NM_001185081.1 |
NP_001172010.1
|
3, 4 |
435 |
|
NM_001185082.1 |
NP_001172011.1
|
3, 4 |
435 |
|
NM_002024.5 |
NP_002015.1
|
3, 4 |
435 |
|
NR_033699.1 |
|
3, 4 |
435 |
|
NR_033700.1 |
|
3, 4 |
435 |
|
GenBank mRNA |
BC038998.1 |
- |
3, 4 |
400 |
|
BC086957.1 |
- |
3, 4 |
465 |
30347992 |
BQ213316.1 |
- |
3, 4 |
387 |
|
KC774027.1 |
- |
3, 4 |
268 |
|
KC774028.1 |
- |
3, 4 |
268 |
|
KC774029.1 |
- |
3, 4 |
268 |
|
KF856234.1 |
- |
3, 4 |
268 |
|
KJ534836.1 |
- |
3, 4 |
206 |
|
KJ534837.1 |
- |
3, 4 |
206 |
|
KJ534838.1 |
- |
3, 4 |
206 |
|
KM091457.1 |
- |
3, 4 |
268 |
|
KM091458.1 |
- |
3, 4 |
268 |
|
KM091459.1 |
- |
3, 4 |
268 |
|
M67468.1 |
- |
3, 4 |
401 |
|
S65791.1 |
- |
3, 4 |
425 |
|
X69962.1 |
- |
3, 4 |
425 |
|
View Full Product Details