Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102603.1 | NP_001096073.1 | 7 | 792 | |
NM_001322128.1 | NP_001309057.1 | 7 | 800 | ||
NM_001322129.1 | NP_001309058.1 | 6 | 895 | ||
NM_001322130.1 | NP_001309059.1 | 7 | 1024 | ||
NM_001322131.1 | NP_001309060.1 | 6 | 899 | ||
NM_001322132.1 | NP_001309061.1 | 7 | 788 | ||
NM_001322133.1 | NP_001309062.1 | 7 | 812 | ||
NM_001322134.1 | NP_001309063.1 | 7 | 816 | ||
NM_001322135.1 | NP_001309064.1 | 6 | 671 | ||
NM_001322136.1 | NP_001309065.1 | 5 | 528 | ||
NM_001322137.1 | NP_001309066.1 | 8 | 848 | ||
NM_001322138.1 | NP_001309067.1 | 7 | 731 | ||
NM_001322139.1 | NP_001309068.1 | 9 | 945 | ||
NM_033288.3 | NP_150630.2 | 6 | 675 | ||
NM_198893.2 | NP_942596.1 | 7 | 760 | ||
XM_017027445.1 | XP_016882934.1 | 4 | 1024 | ||
XM_017027446.1 | XP_016882935.1 | 6 | 985 | ||
GenBank mRNA | AB046831.1 | - | 3 | 1145 | |
AF277623.1 | - | 6 | 683 | ||
AK024442.1 | - | 1 | 39 | ||
BC094880.1 | - | 7 | 677 | 3066532 |