Gene Transcripts

Gene Symbol
RRAGB
Entrez Gene ID
10325
Gene Name
Ras related GTP binding B
Gene Aliases
RAGB, bA465E19.1
Chromosome Location
Chr. X: 55717677 - 55760209 on Build GRCh38
UniGene ID
Hs.50282
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006064.4 NP_006055.3 6, 7 1261
NM_016656.3 NP_057740.2 7, 8 1345
XM_005261973.4 XP_005262030.1 7, 8 682
XM_017029217.1 XP_016884706.1 6, 7 591
XM_017029218.1 XP_016884707.1 7, 8 1301
GenBank mRNA AK289407.1 - 6, 7 1194
AL831926.1 - 6, 7 1200
BC034726.1 - 6, 7 1192 3919252
X90530.1 - 7, 8 1136

View Full Product Details