Gene Transcripts

Gene Symbol
BMPER
Entrez Gene ID
168667
Gene Name
BMP binding endothelial regulator
Gene Aliases
CRIM3, CV-2, CV2
Chromosome Location
Chr. 7: 33904911 - 34155872 on Build GRCh38
UniGene ID
Hs.660998
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133468.4 NP_597725.1 9 1092
XM_005249633.2 XP_005249690.1 8 832
XM_017011800.1 XP_016867289.1 7 652
XM_017011801.1 XP_016867290.1 7 652
GenBank mRNA AK289963.1 - 8 832
AY324650.1 - 9 1091
BC060868.1 - 8 769 30345290

View Full Product Details