Gene Transcripts

Gene Symbol
UPK3A
Entrez Gene ID
7380
Gene Name
uroplakin 3A
Gene Aliases
UP3A, UPIII, UPIIIA, UPK3
Chromosome Location
Chr. 22: 45284987 - 45295874 on Build GRCh38
UniGene ID
Hs.632787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167574.1 NP_001161046.1 4 627
NM_006953.3 NP_008884.1 6 990
GenBank mRNA AF085808.1 - 6 995

View Full Product Details