Gene Transcripts
- Gene Symbol
- RCBTB2
- Entrez Gene ID
- 1102
- Gene Name
- RCC1 and BTB domain containing protein 2
- Gene Aliases
- CHC1L, RLG
- Chromosome Location
- Chr. 13: 48488963 - 48535995 on Build GRCh38
- UniGene ID
- Hs.652712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001268.3 |
NP_001259.1
|
10, 11 |
1376 |
|
NM_001286830.1 |
NP_001273759.1
|
9, 10 |
1311 |
|
NM_001286831.1 |
NP_001273760.1
|
7, 8 |
1116 |
|
XM_005266242.2 |
XP_005266299.1
|
9, 10 |
1229 |
|
XM_005266244.2 |
XP_005266301.1
|
8, 9 |
1164 |
|
XM_006719759.3 |
XP_006719822.1
|
9, 10 |
1197 |
|
XM_011534894.2 |
XP_011533196.1
|
8, 9 |
1124 |
|
XM_017020369.1 |
XP_016875858.1
|
11, 12 |
1650 |
|
XM_017020370.1 |
XP_016875859.1
|
10, 11 |
1522 |
|
XM_017020371.1 |
XP_016875860.1
|
9, 10 |
1395 |
|
XM_017020372.1 |
XP_016875861.1
|
8, 9 |
1005 |
|
XM_017020373.1 |
XP_016875862.1
|
7, 8 |
1187 |
|
GenBank mRNA |
AB209543.1 |
- |
7, 8 |
1082 |
|
AF060219.1 |
- |
9, 10 |
1157 |
|
AK023010.1 |
- |
9, 10 |
1119 |
|
AK301516.1 |
- |
9, 10 |
1135 |
|
AK315703.1 |
- |
10, 11 |
1296 |
|
BC029052.1 |
- |
10, 11 |
1180 |
5194795 |
View Full Product Details