Gene Transcripts

Gene Symbol
Mab21l3
Entrez Gene ID
242125
Gene Name
mab-21-like 3 (C. elegans)
Gene Aliases
Chromosome Location
Chr. 3: 101813076 - 101836286 on Build GRCm38
UniGene ID
Mm.32900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172295.4 NP_758499.3 7 3035
GenBank mRNA AK029040.1 - 7 2774
BB008605.2 - 1 125

View Full Product Details