Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr. 11: 47269376 - 47330031 on Build GRCh38
UniGene ID
Hs.82548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135943.1 NP_001129415.1 4 925
NM_001135944.1 NP_001129416.1 4 901
NM_003682.3 NP_003673.3 4 901
NM_130470.2 NP_569826.2 4 1067
NM_130471.2 NP_569827.2 4 901
NM_130472.2 NP_569828.2 4 901
NM_130473.2 NP_569829.2 4 901
NM_130474.2 NP_569830.2 4 901
NM_130475.2 NP_569831.1 4 925
NM_130476.2 NP_569832.2 4 925
XM_005253189.2 XP_005253246.1 4 925
XM_005253196.2 XP_005253253.1 4 925
XM_005253199.2 XP_005253256.1 4 925
XM_005253200.2 XP_005253257.1 4 925
XM_005253201.2 XP_005253258.1 4 925
XM_005253203.2 XP_005253260.1 4 925
XM_005253204.2 XP_005253261.1 4 925
XM_005253205.1 XP_005253262.1 4 925
XM_017018478.1 XP_016873967.1 4 925
XM_017018479.1 XP_016873968.1 4 925
XM_017018480.1 XP_016873969.1 4 925
XM_017018481.1 XP_016873970.1 4 925
XM_017018482.1 XP_016873971.1 4 925
XM_017018483.1 XP_016873972.1 4 925
XM_017018484.1 XP_016873973.1 4 925
XM_017018485.1 XP_016873974.1 4 886
XM_017018486.1 XP_016873975.1 4 925
XM_017018487.1 XP_016873976.1 4 925
XM_017018488.1 XP_016873977.1 4 925
XM_017018489.1 XP_016873978.1 4 886
XM_017018490.1 XP_016873979.1 4 912
XM_017018491.1 XP_016873980.1 4 881
XM_017018492.1 XP_016873981.1 4 925
XM_017018493.1 XP_016873982.1 4 912
XM_017018494.1 XP_016873983.1 4 925
XM_017018495.1 XP_016873984.1 4 925
XM_017018496.1 XP_016873985.1 4 881
XM_017018497.1 XP_016873986.1 4 886
XM_017018498.1 XP_016873987.1 4 925
XM_017018499.1 XP_016873988.1 4 894
XM_017018500.1 XP_016873989.1 4 925
XM_017018501.1 XP_016873990.1 4 881
XM_017018502.1 XP_016873991.1 4 894
XM_017018503.1 XP_016873992.1 4 881
XM_017018504.1 XP_016873993.1 4 894
XM_017018505.1 XP_016873994.1 4 881
XM_017018506.1 XP_016873995.1 4 886
XM_017018507.1 XP_016873996.1 4 912
XM_017018508.1 XP_016873997.1 4 925
XM_017018509.1 XP_016873998.1 4 881
XM_017018510.1 XP_016873999.1 4 925
GenBank mRNA AB002356.1 - 4 897
AF440100.1 - 4 875
AF440101.1 - 4 875
AF440102.1 - 4 875
AF440103.1 - 4 875
AF440434.1 - 4 875
AK292442.1 - 4 865
BC040484.1 - 4 909 5271798
U44953.1 - 4 889
U77352.1 - 4 1035

View Full Product Details