Gene Transcripts
- Gene Symbol
- Mcpt1
- Entrez Gene ID
- 17224
- Gene Name
- mast cell protease 1
- Gene Aliases
- AV080368, Mcp-1
- Chromosome Location
- Chr. 14: 56017964 - 56020391 on Build GRCm38
- UniGene ID
- Mm.201549
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008570.1 |
NP_032596.1
|
5 |
957 |
|
GenBank mRNA |
S44609.1 |
- |
5 |
950 |
|
View Full Product Details