Gene Transcripts

Gene Symbol
Osbpl1a
Entrez Gene ID
64291
Gene Name
oxysterol binding protein-like 1A
Gene Aliases
G430090F17Rik, Gm753, ORP-1, Osbpl1b
Chromosome Location
Chr. 18: 12755312 - 12941875 on Build GRCm38
UniGene ID
Mm.259470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207530.3 NP_997413.2 7 787
GenBank mRNA AK090069.1 - 7 792
AY536214.1 - 7 666
BC031735.1 - 7 723

View Full Product Details