Gene Transcripts

Gene Symbol
Chmp2b
Entrez Gene ID
68942
Gene Name
charged multivesicular body protein 2B
Gene Aliases
1190006E07Rik
Chromosome Location
Chr. 16: 65539133 - 65562697 on Build GRCm38
UniGene ID
Mm.432944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026879.2 NP_081155.1 6 958
GenBank mRNA AK004506.1 - 3 431
AK084111.1 - 6 960
AK181292.1 - 1 82
AK201890.1 - 1 82
AK206751.1 - 1 82
AK208493.1 - 1 82
AK209621.1 - 1 82
AK215317.1 - 1 82
AK217623.1 - 1 82
AK218468.1 - 1 82
BC042626.1 - 5 714
BC055809.1 - 6 987 6414077

View Full Product Details