Gene Transcripts

Gene Symbol
Itgb3
Entrez Gene ID
29302
Gene Name
integrin, beta 3
Gene Aliases
Chromosome Location
Chr. 10: 92667869 - 92783413 on Build Rnor_6.0
UniGene ID
Rn.229225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153720.1 NP_714942.1 3 223
GenBank mRNA AJ440952.1 - 3 223

View Full Product Details