Gene Transcripts

Gene Symbol
Insig1
Entrez Gene ID
64194
Gene Name
insulin induced gene 1
Gene Aliases
Chromosome Location
Chr. 4: 342302 - 350515 on Build Rnor_6.0
UniGene ID
Rn.772
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022392.1 NP_071787.1 5 987
GenBank mRNA BC078827.1 - 5 997
L13619.1 - 5 987

View Full Product Details