Gene Transcripts

Gene Symbol
Tdo2
Entrez Gene ID
64206
Gene Name
tryptophan 2,3-dioxygenase
Gene Aliases
TO, Tdo
Chromosome Location
Chr. 2: 180897059 - 180914919 on Build Rnor_6.0
UniGene ID
Rn.1029
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022403.2 NP_071798.1 2 125
GenBank mRNA BC089802.1 - 2 121
M55167.1 - 2 125

View Full Product Details