Gene Transcripts
- Gene Symbol
- Idh3a
- Entrez Gene ID
- 114096
- Gene Name
- isocitrate dehydrogenase 3 (NAD+) alpha
- Gene Aliases
- Chromosome Location
- Chr. 8: 59164601 - 59183899 on Build Rnor_6.0
- UniGene ID
- Rn.95104
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053638.1 |
NP_446090.1
|
3, 4 |
239 |
|
GenBank mRNA |
AB047541.1 |
- |
3, 4 |
239 |
|
View Full Product Details