Gene Transcripts
- Gene Symbol
- Pex6
- Entrez Gene ID
- 117265
- Gene Name
- peroxisomal biogenesis factor 6
- Gene Aliases
- Paf-2, Paf2
- Chromosome Location
- Chr. 9: 16568743 - 16580900 on Build Rnor_6.0
- UniGene ID
- Rn.10675
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057125.1 |
NP_476466.1
|
8 |
1876 |
|
GenBank mRNA |
D63673.1 |
- |
8 |
1876 |
|
- Gene Symbol
- LOC100911599
- Entrez Gene ID
- 100911599
- Gene Name
- peroxisome assembly factor 2-like
- Gene Aliases
- Chromosome Location
- Chr. 9: 16317629 - 16323799 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details