Gene Transcripts

Gene Symbol
BCL2L11
Entrez Gene ID
10018
Gene Name
BCL2 like 11
Gene Aliases
BAM, BIM, BOD
Chromosome Location
Chr. 2: 111120914 - 111168445 on Build GRCh38
UniGene ID
Hs.469658
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204106.1 NP_001191035.1 3 447
NM_001204107.1 NP_001191036.1 3 447
NM_001204108.1 NP_001191037.1 3 717
NM_001204110.1 NP_001191039.1 3 447
NM_006538.4 NP_006529.1 4 537
NM_138621.4 NP_619527.1 3 717
NM_138622.3 NP_619528.1 3 717
NM_138623.3 NP_619529.1 4 537
NM_138624.3 NP_619530.1 4 750
NM_138625.3 NP_619531.1 4 563
NM_207003.2 NP_996886.1 3 447
XM_005263550.3 XP_005263607.1 3 1495
XM_005263551.2 XP_005263608.1 3 1495
XM_005263552.3 XP_005263609.1 4 1315
XM_005263553.2 XP_005263610.1 4 1315
XM_005263555.3 XP_005263612.1 3 1225
XM_005263556.3 XP_005263613.1 3 717
XM_005263557.4 XP_005263614.1 3 794
XM_005263559.2 XP_005263616.1 3 1225
XM_005263561.3 XP_005263618.1 4 537
XM_017003102.1 XP_016858591.1 3 447
GenBank mRNA AB071195.1 - 2 429
AB071196.1 - 3 249
AB071197.1 - 3 462
AB071199.1 - 3 275
AF032457.1 - 2 429
AF032458.1 - 3 249
AF455755.1 - 1 129
AK290377.1 - 4 537
AK291269.1 - 3 714
AY305714.1 - 2 159
AY305715.1 - 2 159
AY305716.1 - 2 56
AY423441.1 - 2 159
AY423442.1 - 2 159
AY423443.1 - 2 429
BC033694.1 - 3 699 5213713
DQ849200.1 - 2 159

View Full Product Details