Gene Transcripts
- Gene Symbol
- LINC01549
- Entrez Gene ID
- 100505929
- Gene Name
- long intergenic non-protein coding RNA 1549
- Gene Aliases
- C21orf37
- Chromosome Location
- Chr. 21: 17438890 - 17449185 on Build GRCh38
- UniGene ID
- Hs.46707
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037585.1 |
|
4 |
704 |
|
NR_037586.1 |
|
4 |
698 |
|
GenBank mRNA |
BX089409.1 |
- |
4 |
700 |
|
View Full Product Details