Gene Transcripts

Gene Symbol
Olfr1349
Entrez Gene ID
269862
Gene Name
olfactory receptor 1349
Gene Aliases
MOR232-8
Chromosome Location
Chr. 7: 6514199 - 6520355 on Build GRCm38
UniGene ID
Mm.377869
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207136.1 NP_997019.1 1 848
GenBank mRNA BC147396.1 - 1 870
BC147397.1 - 1 870 9056451

View Full Product Details