Gene Transcripts

Gene Symbol
PTENP1
Entrez Gene ID
11191
Gene Name
phosphatase and tensin homolog pseudogene 1
Gene Aliases
PTEN-rs, PTEN2, PTENpg1, PTH2, psiPTEN
Chromosome Location
Chr. 9: 33673504 - 33677420 on Build GRCh38
UniGene ID
Hs.493716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_023917.1 1 1010
GenBank mRNA AF017999.1 - 1 241
AF019083.1 - 1 241
BC038293.2 - 1 494

View Full Product Details