Gene Transcripts

Gene Symbol
CYP2A7
Entrez Gene ID
1549
Gene Name
cytochrome P450 family 2 subfamily A member 7
Gene Aliases
CPA7, CPAD, CYP2A, CYPIIA7, P450-IIA4
Chromosome Location
Chr. 19: 40875439 - 40885952 on Build GRCh38
UniGene ID
Hs.719890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000764.2 NP_000755.2 7 1668
NM_030589.2 NP_085079.2 6 1515
GenBank mRNA BC117165.1 - 7 1171
BC143325.1 - 7 1171 9051830

View Full Product Details