Gene Transcripts
- Gene Symbol
- ARL13A
- Entrez Gene ID
- 392509
- Gene Name
- ADP ribosylation factor like GTPase 13A
- Gene Aliases
- ARL13, dJ341D10.2
- Chromosome Location
- Chr. X: 100969011 - 100990831 on Build GRCh38
- UniGene ID
- Hs.147237
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103708.1 |
|
8 |
965 |
|
NR_103709.1 |
|
8 |
965 |
|
GenBank mRNA |
AK301813.1 |
- |
8 |
965 |
|
View Full Product Details