Gene Transcripts

Gene Symbol
FREM2
Entrez Gene ID
341640
Gene Name
FRAS1 related extracellular matrix protein 2
Gene Aliases
Chromosome Location
Chr. 13: 38687036 - 38887131 on Build GRCh38
UniGene ID
Hs.253994
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207361.5 NP_997244.4 1 3617
XM_017020554.1 XP_016876043.1 1 3612

View Full Product Details