Gene Transcripts
- Gene Symbol
- CARD8
- Entrez Gene ID
- 22900
- Gene Name
- caspase recruitment domain family member 8
- Gene Aliases
- CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
- Chromosome Location
- Chr. 19: 48203148 - 48256269 on Build GRCh38
- UniGene ID
- Hs.446146
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094933.1 |
- |
5, 6 |
582 |
|
View Full Product Details