Gene Transcripts
- Gene Symbol
- CABLES2
- Entrez Gene ID
- 81928
- Gene Name
- Cdk5 and Abl enzyme substrate 2
- Gene Aliases
- C20orf150, dJ908M14.2, ik3-2
- Chromosome Location
- Chr. 20: 62388630 - 62407283 on Build GRCh38
- UniGene ID
- Hs.301040
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031215.2 |
NP_112492.2
|
10 |
3692 |
|
View Full Product Details