Gene Transcripts

Gene Symbol
CABLES2
Entrez Gene ID
81928
Gene Name
Cdk5 and Abl enzyme substrate 2
Gene Aliases
C20orf150, dJ908M14.2, ik3-2
Chromosome Location
Chr. 20: 62388630 - 62407283 on Build GRCh38
UniGene ID
Hs.301040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031215.2 NP_112492.2 10 3436
GenBank mRNA BC003122.1 - 8 2971

View Full Product Details