Gene Transcripts
- Gene Symbol
- IFNA1
- Entrez Gene ID
- 3439
- Gene Name
- interferon alpha 1
- Gene Aliases
- IFL, IFN, IFN-ALPHA, IFN-alphaD, IFNA13, IFNA@
- Chromosome Location
- Chr. 9: 21440454 - 21441316 on Build GRCh38
- UniGene ID
- Hs.37026
- Species
- Homo sapiens
- Gene Symbol
- IFNA13
- Entrez Gene ID
- 3447
- Gene Name
- interferon alpha 13
- Gene Aliases
- Chromosome Location
- Chr. 9: 21367372 - 21368076 on Build GRCh38
- UniGene ID
- Hs.37026
- Species
- Homo sapiens
- Gene Symbol
- IFNA21
- Entrez Gene ID
- 3452
- Gene Name
- interferon alpha 21
- Gene Aliases
- IFN-alphaI, LeIF F, leIF-F
- Chromosome Location
- Chr. 9: 21165637 - 21166660 on Build GRCh38
- UniGene ID
- Hs.113211
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
V00540.1 |
- |
1 |
435 |
|
View Full Product Details