Gene Transcripts
- Gene Symbol
- RPSA
- Entrez Gene ID
- 3921
- Gene Name
- ribosomal protein SA
- Gene Aliases
- 37LRP, 67LR, ICAS, LAMBR, LAMR1, LBP, LBP/p40, LRP, LRP/LR, NEM/1CHD4, SA, lamR, p40
- Chromosome Location
- Chr. 3: 39406689 - 39412542 on Build GRCh38
- UniGene ID
- Hs.449909
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304288.1 |
NP_001291217.1
|
4, 5 |
629 |
|
NM_002295.5 |
NP_002286.2
|
4, 5 |
614 |
|
GenBank mRNA |
AF503367.1 |
- |
1 |
574 |
|
BC005391.1 |
- |
1 |
586 |
3997019 |
BC008867.2 |
- |
1 |
535 |
4130936 |
BC010418.2 |
- |
1 |
566 |
4079845 |
BC013827.2 |
- |
1 |
570 |
3446816 |
BC018867.2 |
- |
1 |
560 |
|
BC034537.1 |
- |
1 |
567 |
3455545 |
BC050688.1 |
- |
1 |
569 |
4443949 |
BC053370.1 |
- |
1 |
567 |
5108337 |
BC062714.1 |
- |
1 |
590 |
3686924 |
BC066941.1 |
- |
4, 5 |
583 |
4820432 |
BC068062.1 |
- |
1 |
548 |
5427114 |
BC070263.1 |
- |
4, 5 |
614 |
3996864 |
BC071693.1 |
- |
1 |
576 |
3884457 |
BC071968.1 |
- |
1 |
567 |
5814758 |
BC071969.1 |
- |
1 |
555 |
6483856 |
BC071970.1 |
- |
1 |
540 |
6484091 |
BC073863.1 |
- |
1 |
552 |
3924066 |
BC107567.1 |
- |
1 |
587 |
3681016 |
BT007219.1 |
- |
3, 4 |
505 |
|
J03799.1 |
- |
3, 4 |
542 |
|
M14199.1 |
- |
1, 2 |
25 |
|
S37431.1 |
- |
3, 4 |
520 |
|
X15005.1 |
- |
4, 5 |
583 |
|
X61156.1 |
- |
3, 4 |
475 |
|
- Gene Symbol
- FAM149A
- Entrez Gene ID
- 25854
- Gene Name
- family with sequence similarity 149 member A
- Gene Aliases
- MST119, MSTP119
- Chromosome Location
- Chr. 4: 186104798 - 186172663 on Build GRCh38
- UniGene ID
- Hs.744961
- Species
- Homo sapiens
- Gene Symbol
- RPSAP58
- Entrez Gene ID
- 388524
- Gene Name
- ribosomal protein SA pseudogene 58
- Gene Aliases
- RPSA_30_1642
- Chromosome Location
- Chr. 19: 23763014 - 23828135 on Build GRCh38
- UniGene ID
- Hs.560655
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003662.3 |
|
4 |
928 |
|
- Gene Symbol
- RPSAP9
- Entrez Gene ID
- 653162
- Gene Name
- ribosomal protein SA pseudogene 9
- Gene Aliases
- LAMR1P9, RPSA_20_986
- Chromosome Location
- Chr. 9: 76398599 - 76400038 on Build GRCh38
- UniGene ID
- Hs.655646
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026890.1 |
|
1 |
605 |
|
View Full Product Details