Gene Transcripts
- Gene Symbol
- RPSA
- Entrez Gene ID
- 3921
- Gene Name
- ribosomal protein SA
- Gene Aliases
- 37LRP, 67LR, ICAS, LAMBR, LAMR1, LBP, LBP/p40, LRP, LRP/LR, NEM/1CHD4, SA, lamR, p40
- Chromosome Location
- Chr. 3: 39406689 - 39412542 on Build GRCh38
- UniGene ID
- Hs.449909
- Species
- Homo sapiens
- Gene Symbol
- RPSAP58
- Entrez Gene ID
- 388524
- Gene Name
- ribosomal protein SA pseudogene 58
- Gene Aliases
- RPSA_30_1642
- Chromosome Location
- Chr. 19: 23763014 - 23828135 on Build GRCh38
- UniGene ID
- Hs.560655
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003662.3 |
|
4 |
572 |
|
View Full Product Details